Learn More
Invitrogen™ Leptospira biflexa Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA17228
Description
PA1-7228 detects Leptospira biflexa. PA1-7228 has been successfully used in immunofluorescence procedures. The PA1-7228 immunogen is whole Leptospira biflexa lysate. Recommended short term (less than 6 months) storage is liquid at 2-8ºC. For longer term storage, aliquot and freeze.
Leptospirosis is a contagious disease affecting both animals and humans, spread by infection with a bacterial pathogen called Leptospira. Infection may result in chronic liver and kidney disease. Leptospires are known as aquatic spirochetes: they thrive in water and appear long and helical with a characteristic hook on one or both ends. These organisms are divided into two species, Leptospira biflexa and Leptospira interogans.
Specifications
Leptospira biflexa | |
Polyclonal | |
Unconjugated | |
L. biflexa; Leptospira; Leptospirosis | |
Whole Leptospira biflexa lysate. | |
1 mL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Rabbit | |
Protein A | |
RUO | |
Bacteria | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.