Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Leptospira biflexa Polyclonal Antibody, HRP
GREENER_CHOICE

Catalog No. PIPA173009
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA173009 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA173009 Supplier Invitrogen™ Supplier No. PA173009
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The PA1-73009 antibody reacts with Leptospira biflexa. PA1-73009 has been successfully used in ELISA applications. The PA1-73009 antibody was raised against whole L. biflexa organism.

Leptospirosis is a contagious disease affecting both animals and humans, spread by infection with a bacterial pathogen called Leptospira. Infection may result in chronic liver and kidney disease. Leptospires are known as 'aquatic spirochetes': they thrive in water and appear long and helical with a characteristic hook on one or both ends. These organisms are divided into two species, Leptospira biflexa and Leptospira interogans.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Leptospira biflexa
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate HRP
Formulation PBS with 10mg/mL BSA and 0.002% thimerosal
Gene Alias L. biflexa; Leptospira; Leptospirosis
Host Species Rabbit
Immunogen Whole L. biflexa organism.
Purification Method IgG fraction
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.