Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Lestaurtinib
GREENER_CHOICESDP

Catalog No. 33951 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-951 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 33-951 Supplier R&D Systems™ Supplier No. 3395/1

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. JAK2, FLT3 and TrkA inhibitor; also inhibits AurA and AurB

Potent JAK2, FLT3 and TrkA inhibitor (IC50 values are 0.9, 3 and < 25 nM, respectively). Also inhibits Aurora kinase A and B (IC50 values are 8.1 and 2.3 nM, respectively) and prevents STAT5 phosphorylation (IC50 = 20 - 30 nM). Exhibits antiproliferative activity in vitro (IC50 = 30 - 100 nM in HEL92.1.7 cells) and is effective against myeloproliferative disorders in vivo.

Chemical Identifiers

CAS 111358-88-4
Molecular Formula C26H21N3O4

Specifications

Quantity 1 mg
Product Line Tocris
Formula Weight 439.46
Purity >99%
Molecular Weight (g/mol) 439.47
Chemical Name or Material Lestaurtinib
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.