Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LETM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597921
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597921 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597921 Supplier Invitrogen™ Supplier No. PA597921
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LETM1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

This gene encodes a protein that is localized to the inner mitochondrial membrane. The protein functions to maintain the mitochondrial tubular shapes and is required for normal mitochondrial morphology and cellular viability. Mutations in this gene cause Wolf-Hirschhorn syndrome, a complex malformation syndrome caused by the deletion of parts of the distal short arm of chromosome 4. Related pseudogenes have been identified on chromosomes 8, 15 and 19.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LETM1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene LETM1
Gene Accession No. O95202, Q9Z2I0
Gene Alias LETM1; LETM1 and EF-hand domain-containing protein 1, mitochondrial; leucine zipper and EF-hand containing transmembrane protein 1; leucine zipper-EF-hand containing transmembrane protein 1; leucine zipper-EF-hand-containing transmembrane protein 1; Mdm38 homolog; Mitochondrial proton/calcium exchanger protein
Gene Symbols LETM1
Host Species Rabbit
Immunogen Recombinant Human Mitochondrial proton/calcium exchanger protein (443-543AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3954, 56384
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.