Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LETM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543881
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543881 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543881 Supplier Invitrogen™ Supplier No. PA543881
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: KNYESKKYSD PSQPGNTVLH PGTRLIQKLH TSTCWLQEVP GKPQLEQATK Sequence homology: Human: 100%.

LETM2 contains 1 LETM1 domain. The function of the LETM1 protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LETM2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LETM2
Gene Accession No. Q2VYF4
Gene Alias 6030453H13; D030041N04Rik; LETM1 and EF-hand domain-containing protein 2; LETM1 domain-containing protein LETM2, mitochondrial; LETM2; LETM2S; leucine zipper and EF-hand containing transmembrane protein 2; leucine zipper-, EF-hand motif- containing transmembrane protein 2; leucine zipper-, EF-hand motif- containing transmembrane protein 2S; leucine zipper-EF-hand containing transmembrane protein 1-like protein; leucine zipper-EF-hand containing transmembrane protein 2; Leucine zipper-EF-hand-containing transmembrane protein 1-like
Gene Symbols LETM2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human LETM2 (aa 49-98).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 137994
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.