Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LETM2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555209
Description
Immunogen sequence: GLTEEQLRQQ LTEWQDLHLK ENVPPSLLLL SRTFYLIDVK PKPIEIPLSG EAPKTDILVE LPTFTE Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 76%.
LETM2 contains 1 LETM1 domain. The function of the LETM1 protein remains unknown.
Specifications
LETM2 | |
Polyclonal | |
Unconjugated | |
LETM2 | |
6030453H13; D030041N04Rik; LETM1 and EF-hand domain-containing protein 2; LETM1 domain-containing protein LETM2, mitochondrial; LETM2; LETM2S; leucine zipper and EF-hand containing transmembrane protein 2; leucine zipper-, EF-hand motif- containing transmembrane protein 2; leucine zipper-, EF-hand motif- containing transmembrane protein 2S; leucine zipper-EF-hand containing transmembrane protein 1-like protein; leucine zipper-EF-hand containing transmembrane protein 2; Leucine zipper-EF-hand-containing transmembrane protein 1-like | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
137994 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q2VYF4 | |
LETM2 | |
Recombinant protein corresponding to Human LETM2. Recombinant protein control fragment (Product #RP-93882). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction