Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LETM2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | LETM2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
LETM2 Polyclonal specifically detects LETM2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
LETM2 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
137994 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
FLJ25409, LETM1 and EF-hand domain-containing protein 2, LETM1 domain-containing protein LETM2, mitochondrial, leucine zipper-EF-hand containing transmembrane protein 1-like protein, leucine zipper-EF-hand containing transmembrane protein 2, Leucine zipper-EF-hand-containing transmembrane protein 1-like | |
Recombinant fusion protein containing a sequence corresponding to amino acids 257-396 of human LETM2 (NP_653253.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title