Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LETMD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596069
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596069 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596069 Supplier Invitrogen™ Supplier No. PA596069
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FPRQLLIRHF WTPKQQTDFL DIYHAFRKQS HPEIISYLEK VIPLISDAGL RWRLTDLCTK IQRGTHPAIH DILALRECFS NHPLGMNQLQ ALHVKALSRA MLLTSYLPPP LLRHRLKTHT TVIHQLDKAL AKLGIGQLTA QEVKSACYLR GLNSTHIGED RCRTWLGEWL QISCSLKEAE LSLLLHNVVL LSTNYLGTRR; Positive Samples: MCF7, Mouse liver; Cellular Location: Mitochondrion outer membrane, Single-pass membrane protein.

HCCR-1 (human cervical cancer oncogene 1), also known as HCCR-2 or LETMD1, is a 360 amino acid single-pass membrane protein that is expressed in the kidney, liver, skeletal muscle, heart and brain. It is suggested that HCCR-1 may be involved in tumorigenesis and may function as a negative regulator of the p53 tumor suppressor. Overexpression of HCCR-1 may cause mitochondrial dysfunction that can lead to UVC or staurosporine-induced apoptosis resistance and progression of tumor formation. HCCR-1 is considered a candidate biomarker for breast cancer. Various human tumors including leukemia, lymphoma, and carcinomas of the breast, kidney, ovary, stomach, colon, and uterine cervix have high HCCR-1. Six isoforms exist due to alternative splicing events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LETMD1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene LETMD1
Gene Accession No. Q6P1Q0, Q924L1
Gene Alias 1110019O13Rik; AI593524; BB130465; BB235638; Cervical cancer 1 proto-oncogene protein p40; cervical cancer proto-oncogene 2 protein; Cervical cancer receptor; HCCR1; HCCR-1; HCCR-2; HCRR-2; LETM1 domain containing 1; LETM1 domain-containing protein 1; LETMD1; MCC-32; Mccr; regulator of TP53; RGD1310591
Gene Symbols LETMD1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 161-360 of human LETMD1 (NP_0562313).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25875, 681352, 68614
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.