Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Leucomalachite Green Polyclonal Antibody
Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA175233
Description
Working Concentration: 0.156 μg/mL. Sensitivity: 10 ng/mL Leucomalachite Green produces 66% inhibition in a competitive ELISA employing Leucomalachite Green polyclonal antibody.

Specifications
Leucomalachite Green | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.3 | |
Sheep | |
Purified | |
RUO | |
Chemical | |
Antibody | |
IgG |
ELISA | |
Unconjugated | |
C23H26N2; Tetramethyldiaminotriphenylmethane | |
Leucomalachite green-BTG. | |
1 mg | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction