Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Leucyl-cystinyl Aminopeptidase/LNPEP Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP232111AF350
Description
Leucyl-cystinyl Aminopeptidase/LNPEP Polyclonal antibody specifically detects Leucyl-cystinyl Aminopeptidase/LNPEP in Human samples. It is validated for Western BlotSpecifications
Leucyl-cystinyl Aminopeptidase/LNPEP | |
Polyclonal | |
Western Blot | |
angiotensin IV receptor, AT (4) receptor, CAP, Cystinyl aminopeptidase, EC 3.4.11, EC 3.4.11.3, Insulin-regulated membrane aminopeptidase, Insulin-responsive aminopeptidase, IRAP, leucyl/cystinyl aminopeptidase, leucyl-cystinyl aminopeptidase, otase, Oxytocinase, Placental leucine aminopeptidase, PLAP, P-LAPinsulin-regulated aminopeptidase, vasopressinase | |
The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Oxytocinase using the numbering given in entry NP_005566.2 (GeneID 4012). | |
0.1 mL | |
Immunology | |
4012 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction