Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Leukotriene A4 Hydrolase/LTA4H Antibody (455), DyLight 680, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289375FR
Description
Leukotriene A4 Hydrolase/LTA4H Monoclonal antibody specifically detects Leukotriene A4 Hydrolase/LTA4H in Human samples. It is validated for ELISASpecifications
Leukotriene A4 Hydrolase/LTA4H | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
455 | |
ELISA | |
EC 3.3.2.6, Leukotriene A(4) hydrolase, leukotriene A4 hydrolase, leukotriene A-4 hydrolase, LTA4, LTA-4 hydrolase | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Leukotriene A4 Hydrolase/LTA4H (NP_000886.1; Met 1-Asp 611). | |
0.1 mL | |
Lipid and Metabolism | |
4048 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction