Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Leukotriene A4 Hydrolase/LTA4H Antibody (6G7), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32616150UL
Description
Leukotriene A4 Hydrolase/LTA4H Monoclonal antibody specifically detects Leukotriene A4 Hydrolase/LTA4H in Human, Mouse samples. It is validated for Western Blot, ELISASpecifications
Leukotriene A4 Hydrolase/LTA4H | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, ELISA | |
6G7 | |
Western Blot 1:500-1:5000, ELISA | |
EC 3.3.2.6, Leukotriene A(4) hydrolase, leukotriene A4 hydrolase, leukotriene A-4 hydrolase, LTA4, LTA-4 hydrolase | |
A synthesized peptide derived from Human Leukotriene A4 Hydrolase/LTA4H [UniProt P09960] | |
50 μL | |
Lipid and Metabolism | |
4048 | |
Store at -20 to -70C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction