Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Leukotriene B4 R1 Antibody (202/7B1), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064831H
Description
Leukotriene B4 R1 Monoclonal specifically detects Leukotriene B4 R1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
Leukotriene B4 R1 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
LTB4R | |
HeLa cell transfectants expressing human BLTR. | |
0.1 mL | |
GPCR, Immunology, Innate Immunity, Signal Transduction | |
1241 | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Frozen) | |
202/7B1 | |
Immunohistochemistry, Immunohistochemistry-Frozen | |
chemokine receptor-like 1, CMKRL1, leukotriene B4 receptor, LTBR1, P2Y purinoceptor 7, purinergic receptor P2Y, G-protein coupled, 7 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
NB100-64831 recognizes the human BLTR molecule, the leukotriene B4 receptor. BLTR is expressed by peripheral blood granulocytes and is important in the pro-inflammatory response. | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction