Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Leukotriene B4 R1 Antibody (203/14F11), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB099PEATT594
Description
Leukotriene B4 R1 Monoclonal antibody specifically detects Leukotriene B4 R1 in Human samples. It is validated for Flow CytometrySpecifications
Leukotriene B4 R1 | |
Monoclonal | |
PE/ATTO 594 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
203/14F11 | |
Flow Cytometry | |
chemokine receptor-like 1, CMKRL1, leukotriene B4 receptor, LTBR1, P2Y purinoceptor 7, purinergic receptor P2Y, G-protein coupled, 7 | |
HeLa cervical epithelial carcinoma cell line transfected with human Leukotriene B4 R1 | |
0.5 mL | |
GPCR, Immunology, Innate Immunity, Signal Transduction | |
1241 | |
Store at 4C in the dark. Do not freeze. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction