Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Leukotriene B4 R1 Antibody (SR1641), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | Leukotriene B4 R1 |
---|---|
Clone | SR1641 |
Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Description
Leukotriene B4 R1 Monoclonal antibody specifically detects Leukotriene B4 R1 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
Leukotriene B4 R1 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
chemokine receptor-like 1, CMKRL1, leukotriene B4 receptor, LTBR1, P2Y purinoceptor 7, purinergic receptor P2Y, G-protein coupled, 7 | |
A synthesized peptide derived from human Leukotriene B4 R1 (Uniprot #: Q15722) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1641 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
GPCR, Immunology, Innate Immunity, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
1241 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title