Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leupeptin Hemisulfate, PROMO, Thermo Scientific Chemicals

Catalog No. J61188MBPM
Click to view available options
Quantity:
25 mg
100 mg

Reversible protease inhibitor which inhibits cathepsin B, calpain and trypsin

Applications

Reversible protease inhibitor, which inhibits cathepsin B, calpain and trypsinPharmaceutical composition containing leupeptin hemisulfate is used as an anti-malarial agent. It is employed in the treatment of noise-induced hearing loss. It also protects the heart from myocardial stunning. Further, it is used to inhibit serine, cysteine proteases, plasmin, trypsin, papain, kallikrein and cathepsin B.

Solubility

Soluble in water, ethanol, dimethyl formamide and acetic acid.

Notes

Store in a cool place. Incompatible with strong acids and strong bases.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103476-89-7
Molecular Formula C20H39N6O4+

Specifications

Color White
Product Type Leupeptin
Solubility Soluble in water
Content And Storage -30°C to -10°C
Form Powder
Quantity 25 mg

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.