Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Levetiracetam
GREENER_CHOICE

Catalog No. 283910
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-391-0 10 mg
28-395-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 28-391-0 Supplier Tocris Bioscience™ Supplier No. 2839/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Antiepileptic; binds SV2A

Levetiracetam is an antiepileptic that displays distinctive properties from conventional antiepileptic drugs. Displays potent seizure protection in animal models of chronic epilepsy but lacks activity in acute seizure models. Binds synaptic vesicle protein 2A (SV2A) and inhibits Na+-dependent Cl-/HCO3- exchange. In a mouse model of Alzheimer's disease, Levetiracetam rescues levels of presynaptic vesicle fusion and neurotransmitter release, reduces Aβ42 and APP levels and decreases deposits of amyloid plaque; in vitro Levetiracetam inhibits Aβ-induced glutamate from astrocytes.

Chemical Identifiers

CAS 102767-28-2
Molecular Formula C8H14N2O2

Specifications

Quantity 10 mg
Formula Weight 170.21
Purity >99%
Molecular Weight (g/mol) 170.21
Chemical Name or Material Levetiracetam
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.