Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Levodopa Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119575
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119575 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119575 Supplier Invitrogen™ Supplier No. PA5119575
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Levodopa, also known as L-DOPA, is the most widely used drug for the symptomatic treatment of Parkinson's disease, serving as the gold standard for managing the disease's motor symptoms. It functions as a precursor to dopamine, which is deficient in patients with Parkinson's disease due to the degeneration of dopaminergic neurons in the substantia nigra. Once ingested, levodopa crosses the blood-brain barrier and is converted to dopamine, thereby alleviating symptoms such as bradykinesia, rigidity, and tremor. The pharmacokinetics and pharmacodynamics of levodopa have been extensively studied, with various formulations developed to optimize its absorption and reduce side effects. Despite its efficacy, long-term use of levodopa can lead to complications, including motor fluctuations and dyskinesias.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Levodopa
Applications Dot Blot, ELISA
Classification Polyclonal
Concentration 0.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene Alias l-3,4-dihydroxyphenylalanine; L-DOPA
Host Species Rabbit
Immunogen Levodopa (L-DOPA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.