Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Levulinic acid, 98%

Catalog No. p-7081024
Click to view available options
Quantity:
250 g
1000 g

CAS: 123-76-2 | C5H8O3 | 116.116 g/mol

Mainly used as an intermediate for medicines, agricultural chemical and organic synthesis. Used as raw material of perfume, modifier and solvent of plastics. It is also employed as an additive of polymer, paints and lubricating oil. Also used as a surfactant and auxiliary for making printing ink, rubber flux and cosmetics(including shampoo).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mainly used as an intermediate for medicines, agricultural chemical and organic synthesis. Used as raw material of perfume, modifier and solvent of plastics. It is also employed as an additive of polymer, paints and lubricating oil. Also used as a surfactant and auxiliary for making printing ink, rubber flux and cosmetics(including shampoo).

Solubility
Soluble in water ( 675g/L at 20°C).

Notes
Light Sensitive. Store in dark. Store away from oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-76-2
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.116
MDL Number MFCD00002796
InChI Key JOOXCMJARBKPKM-UHFFFAOYSA-N
Synonym levulinic acid, laevulinic acid, pentanoic acid, 4-oxo, levulic acid, 4-oxovaleric acid, 3-acetylpropionic acid, 4-ketovaleric acid, leva, gamma-ketovaleric acid, acetopropionic acid
PubChem CID 11579
ChEBI CHEBI:45630
IUPAC Name 4-oxopentanoic acid
SMILES CC(=O)CCC(=O)O

Specifications

Melting Point 26°C to 31°C
Density 1.134
pH 2.5 to 3.0
Boiling Point 245°C to 246°C
Flash Point 137°C (278°F)
Odor Characteristic
Refractive Index 1.4415
Quantity 250 g
UN Number UN3261
Beilstein 506796
Sensitivity Light sensitive
Merck Index 14,5472
Solubility Information Soluble in water ( 675g/L at 20°C).
Formula Weight 116.12
Percent Purity 98%
Chemical Name or Material Levulinic acid
Show More Show Less
Hazard Category H302-H315-H319
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 204-649-2
RTECSNumber OI1575000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.