Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lewis A Blood Group Antigen Antibody (SPM279), Janelia Fluor™ 646 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311560JF646
Description
Lewis A Blood Group Antigen Monoclonal antibody specifically detects Lewis A Blood Group Antigen in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Lewis A Blood Group Antigen | |
Monoclonal | |
Janelia Fluor 646 | |
A transferase, A3GALNT, A3GALT1, ABO, B transferase, EC=2.4.1.37, EC=2.4.1.40, Fucosylglycoprotein 3-alpha-galactosyltransferase, Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase, Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase soluble form, Glycoprotein-fucosylgalactoside alpha-galactosyltransferase, Glycoprotein-fucosylgalactoside alpha-N-acetylgalactosaminyltransferase, GTB, Histo-blood group A transferase, Histo-blood group ABO system transferase, Histo-blood group B transferase, NAGAT | |
Mucins isolated from ovarian cyst fluid | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
SPM279 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
28 | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction