Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lewis Y Antibody (BR96), Alexa Fluor™ 700, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP281077AF700
Description
Lewis Y Monoclonal specifically detects Lewis Y in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Lewis Y | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
The antibody binds Lewis Y with a Ka of 0.1 uM. | |
Store at 4°C in the dark. | |
IgG κ |
ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
BR96 | |
ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
IPCA-2, IPCA6, IPCA-6, IPCA-8, PCANAP2, PCANAP6, PCANAP8, Prostate cancer-associated protein 6, PRST, Solute carrier family 45 member 3 | |
H3396 cultured line established from a metastasis of a human breast adenocarcinoma. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction