Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lewis Y Antibody (LWY/1463) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$610.00
Specifications
| Antigen | Lewis Y |
|---|---|
| Clone | LWY/1463 |
| Concentration | 1 mg/mL |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, CyTOF-ready |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB254560100
![]() |
Novus Biologicals
NBP254560100UG |
100 μg |
Each for $610.00
|
|
|||||
NB254560200
![]() |
Novus Biologicals
NBP254560200UG |
200 μg | N/A | N/A | N/A | ||||
Description
Lewis Y Monoclonal specifically detects Lewis Y in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Lewis Y | |
| 1 mg/mL | |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| Human colon carcinoma cells | |
| Primary | |
| Store at -20 to -80C. Avoid freeze-thaw cycles. |
| LWY/1463 | |
| Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS with No Preservative | |
| IgG1 κ | |
| Protein A or G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title