Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lewis Y Chimeric Recombinant Rabbit Monoclonal Antibody (BR96)
GREENER_CHOICE

Catalog No. PIMA547983
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547983 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547983 Supplier Invitrogen™ Supplier No. MA547983
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Specificity: The antibody binds Le(Y) with a Ka of 0.1 μM.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Lewis Y Chimeric
Applications ELISA, Immunohistochemistry
Classification Recombinant Monoclonal
Clone BR96
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene BACE2
Gene Accession No. Q9Y5Z0
Gene Alias LleY
Host Species Rabbit
Immunogen H3396 cultured line established from a metastasis of a human breast adenocarcinoma.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25825
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.