Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lewis Y Antibody (LWY/1463), Novus Biologicals™
SDP

Catalog No. p-7078991 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
200 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB25321620 20 μg
NB253216100 100 μg
NB253216200 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB25321620 Supplier Novus Biologicals Supplier No. NBP25321620UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lewis Y Monoclonal specifically detects Lewis Y in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Lewis Y
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone LWY/1463
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 0.5 - 1 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml
Formulation 10mM PBS with 0.05% BSA with 0.05% Sodium Azide
Host Species Mouse
Immunogen Human colon carcinoma cells
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.