Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LFG Monoclonal Antibody (2C2)
GREENER_CHOICE

Catalog No. PIMA521391
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA521391 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA521391 Supplier Invitrogen™ Supplier No. MA521391
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MTQGKLSVAN KAPGTEGQQQ VHGEKKEAPA VPSAPPSYEE ATSGEGMKAG AFPPAPTAVP LHPSWAYVDP SSSSSYDNGF PTGDHELFTT FSWDDQKVRR VFVRKVYTIL LIQLLVTLAV VALFTFCDPV KDYVQANPGW YWASYAVFFA TYLTLACCSG PRRHFPWNLI LLTVFTLSMA YLTGMLSSYY NTTSVLLCLG ITALVCLSVT VFSFQTKFDF TSCQGVLFVL LMTLFFSGLI LAILLPFQYV PWLHAVYAAL GAGVFTLFLA LDTQLLMGNR RHSLSPEEYI FGALNIYLDI IYIFTFFLQL FGTNRE.

LFG, a seven transmembrane spanning protein, is a recently identified member of a family whose members can inhibit apoptotic cell death induced by Fas receptor. LFG is a human homolog of rat NMP35 and consists of a conserved transmembrane domain and a small cytoplasmic domain. Reports have shown that LFG protects the cell from Fas-mediated neuronal cell death in a novel way. LFG binds to Fas receptor but doesn't have any effect in reducing Fas expression or decreasing the ability of FADD binding to the receptor. Furthermore, LFG doesn't protect the cells from TNF-alpha mediated cell death process. Although LFG is ubiquitously expressed, Northern blot analysis detected an increased expression in hippocampus and other neural tissues, suggesting a protective role in neurodegenerative diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LFG
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 2C2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene FAIM2
Gene Accession No. Q9BWQ8
Gene Alias 2900002L20Rik; FAIM2; Fas apoptotic inhibitory molecule 2; fas apoptotic inhibitory molecule 2; protein lifeguard 2; Kiaa0950; Lfg; LFG2; lifeguard; neural membrane protein 35; NGP35; NMP25; Nmp35; protein lifeguard; protein lifeguard 2; TMBIM2; transmembrane BAX inhibitor motif-containing protein 2
Gene Symbols FAIM2
Host Species Mouse
Immunogen FAIM2 (AAH00051, 1 a.a. ~ 316 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23017
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.