Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LFNG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584802
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584802 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584802 Supplier Invitrogen™ Supplier No. PA584802
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TKKFHRARLD LLLETWISRH KEMTFIFTDG EDEALARHTG NVVITNCSAA H Highest antigen sequence indentity to the following orthologs: Rat - 92%, Mouse - 88%.

LFNG belongs to evolutionarily conserved glycosyltransferases that act in the Notch signaling pathway to define boundaries during embryonic development. While their genomic structure is distinct from other glycosyltransferases, fringe proteins have a fucose-specific beta-1,3-N-acetylglucosaminyltransferase activity that leads to elongation of O-linked fucose residues on Notch, which alters Notch signaling. This protein is predicted to be a single-pass type II Golgi membrane protein but it may also be secreted and proteolytically processed like the related proteins in mouse and Drosophila (PMID: 9187150).
TRUSTED_SUSTAINABILITY

Specifications

Antigen LFNG
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LFNG
Gene Accession No. Q8NES3
Gene Alias AW061165; beta-1,3-N-acetylglucosaminyltransferase lunatic fringe; LFNG; LFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase; lunatic fringe; lunatic fringe gene homolog; MGC128664 protein; O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase; SCDO3
Gene Symbols LFNG
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LFNG. Recombinant protein control fragment (Product #RP-103673).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3955
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.