Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LG 101506, Tocris Bioscience™

Catalog No. 350810
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-081-0 10 mg
35-085-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 35-081-0 Supplier R&D Systems Supplier No. 3508/10
This item is not returnable. View return policy

Selective RXR modulator

Selective RXR modulator (Ki values are 3, 9 and 11 nM for RXRα, RXRβ and RXRγ respectively). Displays poor binding affinity for RAR isoforms (Ki values are 2746, 3516 and >10,000 nM for RARα, RARβ and RARγ respectively). Binding to RXR results in selective activation of RXR:PPARγ, RXR:PPARα and RXR:PPARδ heterodimers.

Chemical Identifiers

CAS 331248-11-4
Molecular Formula C25H34F2O3
Molecular Weight (g/mol) 420.541
InChI Key BHIBZAZKKARFIM-UHFFFAOYSA-N
Synonym 7-3,5-di-tert-butyl-2-2,2-difluoroethoxy phenyl-3-methylocta-2,4,6-trienoic acid
PubChem CID 71433850
IUPAC Name 7-[3,5-ditert-butyl-2-(2,2-difluoroethoxy)phenyl]-3-methylocta-2,4,6-trienoic acid
SMILES CC(=CC(=O)O)C=CC=C(C)C1=CC(=CC(=C1OCC(F)F)C(C)(C)C)C(C)(C)C

Specifications

Quantity 10 mg
Formula Weight 420.53
Percent Purity >98%
Chemical Name or Material LG 101506
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.