Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LGALS1 Mouse anti-Human, Mouse, Clone: 6C8.4-1, Abnova™

Catalog No. 89035468 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-468 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-468 Supplier Abnova Corporation Supplier No. MAB14063

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against synthetic peptide of human LGALS1.

  • High specificity
  • High reactivity

Specifications

Antigen LGALS1
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 6C8.4-1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of human LGALS1.
Dilution Flow Cytometry (3-5 ug x 106 cells) Immunocytochemistry Immunofluorescence Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:20) Western Blot The optimal working dilution should be determined by the end user
Formulation In PBS, pH 7.4 (0.09% sodium azide).
Gene Accession No. P09382
Gene Alias DKFZp686E23103/GAL1/GBP
Gene Symbols LGALS1
Host Species Mouse
Immunogen A mixture of two peptides corresponding to internal and C-terminus of human LGALS1.
Purification Method Protein A affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3956
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.