Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LGALS13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119347
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119347 200 μL
PIPA5121797 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119347 Supplier Invitrogen™ Supplier No. PA5119347
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LGALS13 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Lysophospholipases are enzymes that act on biological membranes to regulate the multifunctional lysophospholipids. The protein encoded by this gene has lysophospholipase activity. It is composed of two identical subunits which are held together by disulfide bonds. This protein has structural similarity to several members of the beta-galactoside-binding S-type lectin family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LGALS13
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene LGALS13
Gene Accession No. Q9UHV8
Gene Alias beta-galactoside-binding lectin; GAL13; gal-13; Galactoside-binding soluble lectin 13; galectin 13; galectin-13; lectin, galactoside binding soluble 13; lectin, galactoside-binding, soluble, 13; LGALS13; PLAC8; placental protein 13; placental tissue protein 13; PP13
Gene Symbols LGALS13
Host Species Rabbit
Immunogen Recombinant protein Placental Protein 13. The antigen corresponds to amino acid range 1-139 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29124
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.