Learn More
LGALS3 Mouse anti-Human, Clone: OTI21C1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF804700

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Galactose-specific lectin which binds IgE. May mediate with the alpha-3, beta-1 integrin the stimulation by CSPG4 of endothelial cells migration. Together with DMBT1, required for terminal differentiation of columnar epithelial cells during early embryogenesis. In the nucleus: acts as a pre-mRNA splicing factor. Involved in acute inflammatory responses including neutrophil activation and adhesion, chemoattraction of monocytes macrophages, opsonization of apoptotic neutrophils, and activation of mast cells.Specifications
LGALS3 | |
Monoclonal | |
Unconjugated | |
LGALS3 | |
CBP35, GAL3, GALBP, GALIG, L31, LGALS2, MAC2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
3958 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI21C1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P17931 | |
LGALS3 | |
Full length human recombit protein of human LGALS3 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.