Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LGALS3 Rat anti-Human, Mouse, Clone: M3/38, Abnova™

Catalog No. 89934476 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-934-476 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-934-476 Supplier Abnova Corporation Supplier No. MAB12468

Rat monoclonal antibody raised against native mouse Lgals3.

Specifications

Antigen LGALS3
Applications ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone M3/38
Conjugate Unconjugated
Description Rat monoclonal antibody raised against native mouse Lgals3.
Dilution ELISA Flow Cytometry Immunohistochemistry (Frozen sections) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot The optimal working dilution should be determined by the end user.
Formulation In aqueous buffer (0.09% sodium azide, and may contain carrier protein/stabilizer)
Gene Alias GBP/L-34/Mac-2/gal3
Gene Symbols Lgals3
Host Species Rat
Immunogen Native purifed mouse Lgals3.
Purification Method Affinity purification
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16854
Target Species Human, Mouse
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.