Learn More
LGALS3BP Mouse anti-Human, Clone: OTI1C2, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF503456

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. LGALS3BP has been found elevated in the serum of patients with cancer and in those infected by the human immunodeficiency virus (HIV). It appears to be implicated in immune response associated with natural killer (NK) and lymphokine-activated killer (LAK) cell cytotoxicity. Using fluorescence in situ hybridization the full length 90K cDNA has been localized to chromosome 17q25. The native protein binds specifically to a human macrophage-associated lectin known as Mac-2 and also binds galectin 1.Specifications
LGALS3BP | |
Monoclonal | |
Unconjugated | |
LGALS3BP | |
90K, BTBD17B, CyCAP, M2BP, MAC-2-BP, TANGO10B, gp90 | |
Mouse | |
Affinity Chromatography | |
RUO | |
3959 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI1C2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q08380 | |
LGALS3BP | |
Human recombit protein fragment corresponding to amino acids 19-300 of human LGALS3BP produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.