Learn More
LGALS9 Mouse anti-Human, Clone: OTI16B4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF805652

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. The protein encoded by this gene is an S-type lectin. It is overexpressed in Hodgkin′s disease tissue and might participate in the interaction between the H&RS cells with their surrounding cells and might thus play a role in the pathogenesis of this disease and/or its associated immunodeficiency. Multiple alternatively spliced transcript variants have been found for this gene.Specifications
LGALS9 | |
Monoclonal | |
Unconjugated | |
LGALS9 | |
HUAT, LGALS9A | |
Mouse | |
Affinity Chromatography | |
RUO | |
3965 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI16B4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O00182 | |
LGALS9 | |
Full length human recombit protein of human LGALS9 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.