Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LGALS9C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112996
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112996 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112996 Supplier Invitrogen™ Supplier No. PA5112996
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LGALS9C Polyclonal Antibody for Western Blot, ELISA

This gene was initially thought to represent a pseudogene of galectin 9; however, this transcript has good exon-intron structure and encodes a predicted protein of the same size as and highly similar to galectin 9. This gene is one of two similar loci on chromosome 17p similar to galectin 9 and now thought to be protein-encoding. This gene is the more telomeric gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LGALS9C
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene LGALS9C
Gene Accession No. Q6DKI2
Gene Alias Gal-9C; galectin 9 like; galectin 9C; Galectin-9C; galectin-9-like protein B; lectin, galactoside binding soluble 9C; lectin, galactoside-binding, soluble, 9 (galectin 9) pseudogene; lectin, galactoside-binding, soluble, 9C; LGALS9C
Gene Symbols LGALS9C
Host Species Rabbit
Immunogen Recombinant Human Galectin-9C protein (1-356AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 654346
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.