Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LGALSL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570064
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570064 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570064 Supplier Thermo Scientific Supplier No. PA570064
Add to Cart
Add to Cart

LGALSL Polyclonal Antibody

LGALSL Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen LGALSL
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene LGALSL
Gene Accession No. Q8VED9
Gene Alias 1110067D22Rik; A530071M23; galectin like; Galectin-like protein; Galectin-related protein; galectin-related protein A; GRP; Grpa; HSPC159; Lectin galactoside-binding-like protein; lectin galactoside-binding-like protein A; lectin, galactoside binding like; lectin, galactoside binding-like; lectin, galactoside-binding-like; Lgalsl; Lgalsla; RGD1307414
Gene Symbols LGALSL
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human LGALSL (aa 73-122).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 216551
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.