Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LGALSL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590100
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590100 Supplier Invitrogen™ Supplier No. PA590100
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAGSVADSDA VVKLDDGHLN NSLSSPVQAD VYFPRLIVPF CGHIKGGMRP GKKVLVMGIV DLNPESFAIS LTCGDSEDPP ADVAIELKAV FTDRQLLRNS CISGERGEEQ SAIPYFPFIP DQPFRVEILC EHPRFRVFVD GHQLFDFYHR IQTLSAIDTI KINGDLQITK LG; Positive Samples: Mouse brain.

Does not bind lactose, and may not bind carbohydrates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LGALSL
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene LGALSL
Gene Accession No. Q8VED9
Gene Alias 1110067D22Rik; A530071M23; galectin like; Galectin-like protein; Galectin-related protein; galectin-related protein A; GRP; Grpa; HSPC159; Lectin galactoside-binding-like protein; lectin galactoside-binding-like protein A; lectin, galactoside binding like; lectin, galactoside binding-like; lectin, galactoside-binding-like; Lgalsl; Lgalsla; RGD1307414
Gene Symbols LGALSL
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-172 of human LGALSL (NP_0549002).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 216551
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.