Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LGALSL Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA561143
Description
Immunogen sequence: LNNSLSSPVQ ADVYFPRLIV PFCGHIKGGM RPGKKVLVMG IVDLNPESFA ISLTCGDSED PPADVAIELK Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
Does not bind lactose, and may not bind carbohydrates.
Specifications
LGALSL | |
Polyclonal | |
Unconjugated | |
LGALSL | |
1110067D22Rik; A530071M23; galectin like; Galectin-like protein; Galectin-related protein; galectin-related protein A; GRP; Grpa; HSPC159; Lectin galactoside-binding-like protein; lectin galactoside-binding-like protein A; lectin, galactoside binding like; lectin, galactoside binding-like; lectin, galactoside-binding-like; Lgalsl; Lgalsla; RGD1307414 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29094 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q3ZCW2 | |
LGALSL | |
Recombinant protein corresponding to Human LGALSL | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction