Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lgi1 Antibody (S283-7), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242190F
Description
Lgi1 Monoclonal specifically detects Lgi1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Lgi1 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
LGI1 | |
Fusion protein amino acids 37-113 (LRRNT domain and first LRR repeat) of mouse LGI1. Rat: 100% identity (77/77 amino acids identical). Human: 98% identity (76/77 amino acids identical). ∼50% identity with LGI2, LGI3 and LGI4. | |
0.1 mL | |
Primary | |
approx 50% identity with LGI2, LGI3 and LGI4.Detects approx 60kDa. | |
Store at 4C in the dark. | |
IgG2a |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
S283-7 | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
epilepsy, partial, EPITEMPIN, Epitempin-1, EPTleucine-rich glioma-inactivated protein 1, ETL1epitempin-1, IB1099, leucine-rich, glioma inactivated 1 | |
Mouse | |
Protein G purified | |
Neuroscience | |
9211 | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction