Learn More
Invitrogen™ LGI1 Monoclonal Antibody (N283/7), PerCP

Description
Rat: 100% identity (77/77 amino acids identical). Human: 98% identity (76/77 amino acids identical). ~50% identity with LGI2, LGI3 and LGI4. 1 μg/mL of MA5-45679 was sufficient for detection of LGI1 in 20 μg of rat brain lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 60kDa. This antibody was formerly sold as clone S283-7.
Specifications
Specifications
| Antigen | LGI1 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | N283/7 |
| Concentration | 1 mg/mL |
| Conjugate | PerCP |
| Formulation | 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4 |
| Gene | LGI1 |
| Gene Accession No. | O95970, Q8K4Y5, Q9JIA1 |
| Gene Alias | ADLTE; ADPAEF; ADPEAF; BB130740; EPITEMPIN; Epitempin 1; Epitempin-1; EPT; ETL1; IB1099; leucine rich glioma inactivated 1; leucine-rich glioma inactivated 1; leucine-rich glioma-inactivated protein 1; leucine-rich repeat LGI family, member 1; leucine-rich, glioma inactivated 1; LGI1; OTTHUMP00000020121; OTTHUMP00000020122; UNQ775/PRO1569 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.