Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lgi1 Mouse anti-Human, Mouse, Rat, APC, Clone:S283-7, Abnova™

Catalog No. 89333754 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-754 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-754 Supplier Abnova Corporation Supplier No. MAB17083
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant mouse Lgi1.

Specifications

Antigen Lgi1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone S283-7
Conjugate APC
Description leucine-rich repeat LGI family, member 1
Dilution Immunocytochemistry
Immunofluorescence
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections)
Western Blot (1:1000)
The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene Lgi1
Gene Alias BB130740
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 37-113 at LRRNT domain and first LRR repeat of mouse Lgi1.
Quantity 100 μg
Gene ID (Entrez) 56839
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.