Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LGI3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981748
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1748 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1748 Supplier Bioss Supplier No. BS11879R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The leucine-rich, glioma inactivated gene 3 (LGI3) is a member of the LGI family in which LGI1 is the exemplar. The LGI family consists of four of highly related proteins containing leucine-rich repeats (LRRs) which are highly similar to other transmembrane signaling molecules and receptors. LGI1 has been identified as a candidate tumor suppressor gene for glioma and plays a role in autodominant lateral temporal epilepsy (ADTLE), an epileptic syndrome characterized by focal seizures with predominant auditory symptoms. Despite its high homology with LGI1 and similar pattern of expression, mutations in LGI3 have not been found to be associated with ADTLE. LGI3 expression is induced in rat astrocyte cultures by the amyloid beta (Abeta) peptide and accumulated on neuronal plasma membranes of aged monkey brains and co-localized with Abeta.

Specifications

Antigen LGI3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Lgi3
Gene Accession No. Q8N145
Gene Alias AI841179; AW049851; Leubrin; leucine rich repeat LGI family member 3; Leucine-rich glioma-inactivated protein 3; Leucine-rich repeat LGI family member 3; leucine-rich repeat LGI family, member 3; LGI1-like protein 4; LGI3; LGIL4; UNQ8190/PRO23199
Gene Symbols Lgi3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human LGI3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 203190
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.