Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LGI4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560416
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560416 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560416 Supplier Invitrogen™ Supplier No. PA560416
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SMFRLLQQLP SRGAHVFQPL LIARDQLAIL GSDFAFSQVL RLEPDKGLLE PLQELGPPAL VAPRAFAHIT MAGRRFLFAA CFKGPTQIYQ HHEIDLSA Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 90%.

The leucine-rich, glioma inactivated gene 4 (LGI4) is a member of the LGI family in which LGI1 is the exemplar. The LGI family consists of four of highly related proteins containing leucine-rich repeats (LRRs) which are highly similar to other transmembrane signaling molecules and receptors. LGI1 has been identified as a candidate tumor suppressor gene for glioma and plays a role in autodominant lateral temporal epilepsy (ADTLE), an epileptic syndrome characterized by focal seizures with predominant auditory symptoms. Despite its high homology with LGI1 and similar pattern of expression, mutations in LGI4 have not been found to be associated with ADTLE. However, the LGI4 gene is located in a region linked to benign familial infantile convulsions. Further study revealed that a GC-to-AT polymorphism was correlated with childhood absence epilepsy. Other studies showed that decreasing LGI4 expression in cultured cells inhibits myelination, indicating that LGI4 may play a role in neural development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LGI4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LGI4
Gene Accession No. Q8N135
Gene Alias clp; leucine rich repeat LGI family member 4; leucine-rich glioma inactivated protein 4; leucine-rich glioma-inactivated gene 4; leucine-rich glioma-inactivated protein 4; Leucine-rich repeat LGI family member 4; leucine-rich repeat LGI family, member 4; LGI1-like protein 3; LGI4; Lgil3; UNQ6515/PRO21485
Gene Symbols LGI4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LGI4. Recombinant protein control fragment (Product #RP-99116).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 163175
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.