Learn More
Invitrogen™ LGI4 Polyclonal Antibody

Description
A suggested positive control is rat brain tissue lysate. PA5-20515 can be used with blocking peptide PEP-0635.
Specifications
Specifications
| Antigen | LGI4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | LGI4 |
| Gene Accession No. | Q8K1S1, Q8N135 |
| Gene Alias | clp; leucine rich repeat LGI family member 4; leucine-rich glioma inactivated protein 4; leucine-rich glioma-inactivated gene 4; leucine-rich glioma-inactivated protein 4; Leucine-rich repeat LGI family member 4; leucine-rich repeat LGI family, member 4; LGI1-like protein 3; LGI4; Lgil3; UNQ6515/PRO21485 |
| Gene Symbols | LGI4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.