Learn More
Invitrogen™ LGMN Monoclonal Antibody (312114)

Description
In direct ELISAs, no cross-reactivity with recombinant mouse Legumain is observed. This antibody recognizes the pro form of human Legumain and does not recognize the mature form (aa 18-323). Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | LGMN |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 312114 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | LGMN |
| Gene Accession No. | Q99538 |
| Gene Alias | AEP; AI746452; Asparaginyl endopeptidase; AU022324; cb173; cb460; cysteine protease 1; legumain; Lgmn; LGMN1; preprolegumain; protease, cysteine 1; protease, cysteine, 1; protease, cysteine, 1 (legumain); PRSC1; PRSC1EC 3.4.22.34; sb:cb173; wu:fa98g05; wu:fj92g08; zgc:76953 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.