Learn More
LGN Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303032AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
The protein encoded by this gene belongs to a family of proteins that modulate activation of G proteins, which transduce extracellular signals received by cell surface receptors into integrated cellular responses. The N-terminal half of this protein contains 10 copies of leu-gly-asn repeat, and the C-terminal half contains 4 GoLoco motifs, which are involved in guanine nucleotide exchange. This protein may play a role in neuroblast division and in the development of normal hearing. Mutations in this gene are associated with autosomal recessive nonsyndromic deafness .Specifications
LGN | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P81274 | |
GPSM2 | |
Between 627 and 677 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
GPSM2 | |
GPSM2; G-protein signalling modulator 2 (AGS3-like, C. elegans) ; HGNC:29501; LGN; Pins; LGN protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29899 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.