Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LGP2 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518783
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
Anti-viral innate immune responses are triggered by pathogen-associated molecular patters (such as the accumulation of intracellular nucleic acids resulting from virus infections) and represent the first line of defense against numerous infectious organisms. The Toll-like receptors (TLR) 3, 7, 8, and 9 are expressed in immune cells and function as transmembrane pattern recognition receptors to detect foreign nucleic acids. Other proteins that play similar roles, such as RIG-1 and MDA5 are members of a CARD-helicase family and are expressed in the cytoplasm. A third protein, LGP2, is similar to RIG-1 and MDA5, except for lacking a homologous CARD domain. It is thought to act as an element of negative-feedback regulation of intracellular antiviral signaling. At least four isoforms of LGP2 are known to exist.
Specifications
| LGP2 | |
| Polyclonal | |
| Unconjugated | |
| DHX58 | |
| B430001I08Rik; D11LGP2; D11lgp2e; DEXH (Asp-Glu-X-His) box polypeptide 58; DEXH-box helicase 58; DHX58; Lgp2; LPG2; ortholog of mouse D11lgp2; probable ATP-dependent helicase LGP2; Probable ATP-dependent RNA helicase DHX58; protein D11Lgp2; Protein D11Lgp2 homolog; RGD1310093; RIG-I-like receptor 3; RIG-I-like receptor Lgp2; RLR; RLR-3; RNA helicase LGP2 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 79132 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q96C10 | |
| DHX58 | |
| Synthetic peptide sequence (DFLQHCAENLSD) corresponding to the C-terminus amino acids of DHX58 (aa 663-675). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction