Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lgr4/GPR48 Antibody (OTI2D1), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272006AF594
Description
Lgr4/GPR48 Monoclonal specifically detects Lgr4/GPR48 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.Specifications
Lgr4/GPR48 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
LGR4 | |
Recombinant protein fragment corresponding to amino acids 230-540 of human Lgr4/GPR48 (NP_060960 ) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Flow Cytometry | |
OTI2D1 | |
Western Blot, Flow Cytometry | |
GPR48G protein-coupled receptor 48, G-protein coupled receptor 48, leucine-rich repeat containing G protein-coupled receptor 4 | |
Mouse | |
Affinity Purified | |
GPCR | |
55366.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction