Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lgr4/GPR48 Antibody (OTI2D1), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP200974
Description
Lgr4/GPR48 Monoclonal specifically detects Lgr4/GPR48 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.Specifications
Lgr4/GPR48 | |
Monoclonal | |
0.73 mg/mL | |
Western Blot 1:500, Flow Cytometry 1:100 | |
GPR48G protein-coupled receptor 48, G-protein coupled receptor 48, leucine-rich repeat containing G protein-coupled receptor 4 | |
Mouse | |
104 kDa | |
0.1 mL | |
GPCR | |
55366 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Flow Cytometry | |
OTI2D1 | |
Unconjugated | |
Q9BXB1 | |
LGR4 | |
Human recombinant protein fragment corresponding to amino acids 230-540 of human LGR4 (NP_060960 ) produced in HEK293T Cell. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction