Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LGR4 Monoclonal Antibody (OTI5B8), TrueMAB™, OriGene
SDP

Catalog No. 502541047
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1047 30 μL
50-167-4165 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1047 Supplier OriGene Supplier No. TA503215S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LGR4 Monoclonal antibody specifically detects LGR4 in Canine, Human samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen LGR4
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI5B8
Concentration 0.78 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ACY1
Gene Accession No. Q03154
Gene Alias 9130225G07; A330106J01Rik; A930009A08Rik; BNMD17; G protein-coupled receptor 48; G protein-coupled receptor GPR48; GPR48; G-protein coupled receptor 48; leucine rich repeat containing G protein-coupled receptor 4; leucine-rich repeat containing G protein-coupled receptor 4; leucine-rich repeat-containing G protein-coupled receptor 4; leucine-rich repeat-containing G-protein coupled receptor 4; Lgr4
Host Species Mouse
Immunogen Full length human recombinant protein of human ACY1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 476602, 95
Target Species Canine, Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.