Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LGR4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533887
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533887 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533887 Supplier Invitrogen™ Supplier No. PA533887
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Elephant, Panda, Bovine, Dog, Horse, Rabbit, Pig (100%).

G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins. GPCR LGR4 is a member of this family (subfamily Orphan-A). Its expression has been documented in human brain, adrenal, heart, kidney, liver, skeletal muscle, ovary, pancreas, placenta, prostate, spleen, stomach, testis, thyroid, and spinal cord.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LGR4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene LGR4
Gene Accession No. F1MLX5, Q9BXB1
Gene Alias 9130225G07; A330106J01Rik; A930009A08Rik; BNMD17; G protein-coupled receptor 48; G protein-coupled receptor GPR48; GPR48; G-protein coupled receptor 48; leucine rich repeat containing G protein-coupled receptor 4; leucine-rich repeat containing G protein-coupled receptor 4; leucine-rich repeat-containing G protein-coupled receptor 4; leucine-rich repeat-containing G-protein coupled receptor 4; Lgr4
Gene Symbols LGR4
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from C-terminus of human LGR4.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100072305, 100356429, 100518646, 476896, 505423, 55366
Target Species Human, Canine, Rabbit, Monkey, Bovine, Pig, Horse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.